Gene Transcripts

Gene Symbol
Ifna13
Entrez Gene ID
230396
Gene Name
interferon alpha 13
Gene Aliases
Chromosome Location
Chr. 4: 88643641 - 88644459 on Build GRCm38
UniGene ID
Mm.246592
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177347.2 NP_796321.1 1 643
GenBank mRNA AY190047.1 - 1 643
BC120724.1 - 1 599 40129647
BC120750.1 - 1 599 40129673

View Full Product Details