Gene Transcripts

Gene Symbol
Atmin
Entrez Gene ID
234776
Gene Name
ATM interactor
Gene Aliases
Asciz, gpg6, mKIAA0431
Chromosome Location
Chr. 8: 116943386 - 116960445 on Build GRCm38
UniGene ID
Mm.259736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177700.4 NP_808368.3 4 1428
GenBank mRNA AK087334.1 - 1 739
AK122281.1 - 4 1310
AX775769.1 - 4 1428
BC025922.1 - 1 34
BC058974.1 - 3 973
BC060631.1 - 4 1413 6847850

View Full Product Details