Gene Transcripts

Gene Symbol
Apol10b
Entrez Gene ID
328561
Gene Name
apolipoprotein L 10B
Gene Aliases
9130218O11Rik
Chromosome Location
Chr. 15: 77584106 - 77598542 on Build GRCm38
UniGene ID
Mm.379486
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177820.4 NP_808488.2 4 922
GenBank mRNA AK033678.1 - 894
AK144113.1 - 4 926
BC147348.1 - 868 9056388
BC147349.1 - 868 9056408

View Full Product Details