Gene Transcripts

Gene Symbol
Creb3l2
Entrez Gene ID
208647
Gene Name
cAMP responsive element binding protein 3-like 2
Gene Aliases
BBF2H7, C530025K05Rik, SCIRR69
Chromosome Location
Chr. 6: 37327255 - 37442186 on Build GRCm38
UniGene ID
Mm.169929
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178661.4 NP_848776.2 12 3250
GenBank mRNA AK036712.1 - 12 3242
AK082964.1 - 1 198
AK086719.1 - 1 556
AK086757.1 - 1 556

View Full Product Details