Gene Transcripts

Gene Symbol
Atcay
Entrez Gene ID
16467
Gene Name
ataxia, cerebellar, Cayman type homolog (human)
Gene Aliases
3322401A10Rik, BB077577, BNIP-H, CLAC, hes, ji
Chromosome Location
Chr. 10: 81204513 - 81230779 on Build GRCm38
UniGene ID
Mm.127681
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178662.3 NP_848777.1 13 3491
GenBank mRNA AK038990.1 - 13 3492
BC048903.1 - 13 3475 6491141

View Full Product Details