Gene Transcripts

Gene Symbol
AP2A1
Entrez Gene ID
160
Gene Name
adaptor related protein complex 2 alpha 1 subunit
Gene Aliases
ADTAA, AP2-ALPHA, CLAPA1
Chromosome Location
Chr. 19: 49766923 - 49807114 on Build GRCh38
UniGene ID
Hs.467125
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014203.2 NP_055018.2 18, 19 2537
NM_130787.2 NP_570603.2 17, 18 2471
XM_011526556.2 XP_011524858.1 18, 19 3147
XM_011526557.2 XP_011524859.1 17, 18 3076
GenBank mRNA AK094317.1 - 8, 9 973
AL136925.1 - 17, 18 2337
BC014214.2 - 17, 18 2369 4111165

View Full Product Details