Gene Transcripts
- Gene Symbol
- AGXT
- Entrez Gene ID
- 189
- Gene Name
- alanine-glyoxylate aminotransferase
- Gene Aliases
- AGT, AGT1, AGXT1, PH1, SPAT, SPT, TLH6
- Chromosome Location
- Chr. 2: 240868745 - 240879119 on Build GRCh38
- UniGene ID
- Hs.144567
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000030.2 |
NP_000021.1
|
1, 2 |
274 |
|
GenBank mRNA |
AB292648.1 |
- |
1, 2 |
175 |
|
AF348451.1 |
- |
1, 2 |
65 |
|
AK292754.1 |
- |
1, 2 |
195 |
|
AK298400.1 |
- |
1, 2 |
60 |
|
AK298420.1 |
- |
1, 2 |
190 |
|
AK315331.1 |
- |
1, 2 |
222 |
|
AK316495.1 |
- |
1, 2 |
60 |
|
BC132819.1 |
- |
1, 2 |
271 |
40146841 |
D13368.1 |
- |
1, 2 |
261 |
|
X53414.1 |
- |
1, 2 |
275 |
|
X56092.1 |
- |
1, 2 |
174 |
|
View Full Product Details