Gene Transcripts

Gene Symbol
BRDT
Entrez Gene ID
676
Gene Name
bromodomain testis associated
Gene Aliases
BRD6, CT9
Chromosome Location
Chr. 1: 91949371 - 92014428 on Build GRCh38
UniGene ID
Hs.482520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242805.2 NP_001229734.2 6 947
NM_001242806.2 NP_001229735.2 5 890
NM_001242807.2 NP_001229736.2 4 546
NM_001242808.2 NP_001229737.2 4 740
NM_001242810.2 NP_001229739.2 5 1234
NM_001726.4 NP_001717.3 5 673
NM_207189.3 NP_997072.2 5 878
XM_006710853.3 XP_006710916.1 6 1463
XM_006710854.3 XP_006710917.1 7 1308
XM_006710855.3 XP_006710918.1 7 1313
XM_006710856.3 XP_006710919.1 7 1287
XM_006710857.3 XP_006710920.1 7 1277
XM_011542032.2 XP_011540334.1 7 797
XM_011542033.2 XP_011540335.1 7 771
XM_011542034.2 XP_011540336.1 7 787
XM_011542035.2 XP_011540337.1 7 761
XM_011542036.2 XP_011540338.1 5 676
GenBank mRNA AF019085.1 - 5 636
AK302758.1 - 4 482
AK303008.1 - 5 638
AK316442.1 - 4 545
AY338951.1 - 6 947
BC005281.1 - 5 604
BC017582.1 - 5 679
BC047900.1 - 5 643
BC062700.1 - 5 638
BG717291.1 - 5 670
BX648135.1 - 5 969

View Full Product Details