Gene Transcripts

Gene Symbol
CD2
Entrez Gene ID
914
Gene Name
CD2 molecule
Gene Aliases
LFA-2, SRBC, T11
Chromosome Location
Chr. 1: 116754464 - 116769229 on Build GRCh38
UniGene ID
Hs.523500
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK223393.1 - 5 900
AK313614.1 - 5 929
BC033583.1 - 5 884 5227138
M14362.1 - 5 880
M16336.1 - 5 894
M16445.1 - 5 877

View Full Product Details