Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000397.3 | NP_000388.2 | 11 | 1499 | |
GenBank mRNA | AK289753.1 | - | 11 | 1484 | |
AK301476.1 | - | 9 | 1106 | ||
AK301491.1 | - | 10 | 1361 | ||
AK312929.1 | - | 11 | 1460 | ||
BC032720.1 | - | 11 | 1485 | 5505170 | |
KU178009.1 | - | 11 | 1438 | ||
X04011.1 | - | 11 | 1453 |