Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318503.1 | NP_001305432.1 | 4 | 1167 | |
NM_001395.3 | NP_001386.1 | 4 | 1113 | ||
XM_011531123.1 | XP_011529425.1 | 4 | 1026 | ||
XM_011531124.1 | XP_011529426.1 | 4 | 1159 | ||
GenBank mRNA | AK314250.1 | - | 4 | 1113 | |
BC034936.1 | - | 4 | 821 | ||
BC042166.1 | - | 4 | 821 | ||
BC060837.1 | - | 4 | 1167 | 30339218 | |
U87167.1 | - | 2 | 109 | ||
Y08302.1 | - | 4 | 1022 |