Gene Transcripts
- Gene Symbol
- GRIN2B
- Entrez Gene ID
- 2904
- Gene Name
- glutamate ionotropic receptor NMDA type subunit 2B
- Gene Aliases
- EIEE27, GluN2B, MRD6, NMDAR2B, NR2B, hNR3
- Chromosome Location
- Chr. 12: 13537337 - 13982012 on Build GRCh38
- UniGene ID
- Hs.504844
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000834.3 |
NP_000825.2
|
11, 12 |
2549 |
|
XM_005253351.3 |
XP_005253408.1
|
2, 3 |
276 |
|
XM_011520628.2 |
XP_011518930.1
|
12, 13 |
3342 |
|
XM_011520629.2 |
XP_011518931.1
|
13, 14 |
3149 |
|
XM_017019219.1 |
XP_016874708.1
|
12, 13 |
2897 |
|
GenBank mRNA |
AB208850.1 |
- |
8, 9 |
1328 |
|
BC113618.1 |
- |
11, 12 |
2448 |
8322670 |
BC113620.1 |
- |
11, 12 |
2448 |
8322672 |
U11287.1 |
- |
11, 12 |
2580 |
|
U28758.1 |
- |
1, 2 |
90 |
|
U28862.1 |
- |
3, 4 |
390 |
|
U88963.1 |
- |
11, 12 |
2549 |
|
U90278.1 |
- |
11, 12 |
2579 |
|
View Full Product Details