Gene Transcripts

Gene Symbol
MSI1
Entrez Gene ID
4440
Gene Name
musashi RNA binding protein 1
Gene Aliases
Chromosome Location
Chr. 12: 120339655 - 120369180 on Build GRCh38
UniGene ID
Hs.158311
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002442.3 NP_002433.1 2, 3 177
XM_006719403.3 XP_006719466.1 2, 3 171
XM_011538361.2 XP_011536663.1 2, 3 171
XM_011538362.2 XP_011536664.1 2, 3 171
XM_011538363.2 XP_011536665.1 2, 3 171
GenBank mRNA AB012851.1 - 2, 3 152
BX393884.2 - 2, 3 177

View Full Product Details