Gene Transcripts
- Gene Symbol
- SEPT5
- Entrez Gene ID
- 5413
- Gene Name
- septin 5
- Gene Aliases
- CDCREL, CDCREL-1, CDCREL1, H5, HCDCREL-1, PNUTL1
- Chromosome Location
- Chr. 22: 19714464 - 19723322 on Build GRCh38
- UniGene ID
- Hs.728762
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001009939.2 |
NP_001009939.1
|
10, 11 |
1369 |
|
NM_002688.5 |
NP_002679.2
|
11, 12 |
1175 |
|
GenBank mRNA |
AK056273.1 |
- |
10, 11 |
1346 |
|
AK295940.1 |
- |
11, 12 |
1044 |
|
BC025261.1 |
- |
11, 12 |
1139 |
4622405 |
CR456545.1 |
- |
11, 12 |
1055 |
|
KJ534756.1 |
- |
11, 12 |
1050 |
|
KJ534757.1 |
- |
6, 7 |
315 |
|
U74628.1 |
- |
11, 12 |
1137 |
|
Y11593.1 |
- |
11, 12 |
1095 |
|
- Gene Symbol
- SEPT5-GP1BB
- Entrez Gene ID
- 100526833
- Gene Name
- SEPT5-GP1BB readthrough
- Gene Aliases
- Chromosome Location
- Chr. 22: 19717220 - 19724774 on Build GRCh38
- UniGene ID
- Hs.283743
- Species
- Homo sapiens
View Full Product Details