Gene Transcripts

Gene Symbol
REEP5
Entrez Gene ID
7905
Gene Name
receptor accessory protein 5
Gene Aliases
C5orf18, D5S346, DP1, TB2, YOP1, Yip2e
Chromosome Location
Chr. 5: 112876384 - 112922334 on Build GRCh38
UniGene ID
Hs.429608
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005669.4 NP_005660.4 1, 2 255
XM_017009844.1 XP_016865333.1 1, 2 172
GenBank mRNA AK295436.1 - 1, 2 146
AK298268.1 - 1, 2 159
AK299902.1 - 1, 2 151
AK300048.1 - 1, 2 160
AK312522.1 - 1, 2 160
AY562243.1 - 1, 2 111
BC000232.1 - 1, 2 136 3350749
BC065926.1 - 1, 2 114 6164621
BX648005.1 - 1, 2 236
DA807043.1 - 1, 2 256
M74090.1 - 1, 2 137

View Full Product Details