Gene Transcripts

Gene Symbol
ALDH5A1
Entrez Gene ID
7915
Gene Name
aldehyde dehydrogenase 5 family member A1
Gene Aliases
SSADH, SSDH
Chromosome Location
Chr. 6: 24494969 - 24537207 on Build GRCh38
UniGene ID
Hs.371723
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080.3 NP_001071.1 8 1214
NM_170740.1 NP_733936.1 9 1253
GenBank mRNA AJ427354.2 - 9 1021
AJ427355.2 - 8 1186
AK290800.1 - 8 1332
AK294699.1 - 9 991
AK314357.1 - 8 1214
AK315380.1 - 8 1309
BC034321.1 - 8 1214 4829533
KJ534766.1 - 8 1129
KJ534767.1 - 8 1186
KJ534768.1 - 8 1186
KJ534769.1 - 9 1225
L34820.1 - 5 550
Y11192.1 - 8 1186

View Full Product Details