Gene Transcripts
- Gene Symbol
- ADAM21
- Entrez Gene ID
- 8747
- Gene Name
- ADAM metallopeptidase domain 21
- Gene Aliases
- ADAM 21, ADAM31
- Chromosome Location
- Chr. 14: 70452157 - 70459905 on Build GRCh38
- UniGene ID
- Hs.178748
- Species
- Homo sapiens
- Gene Symbol
- ADAM21P1
- Entrez Gene ID
- 145241
- Gene Name
- ADAM metallopeptidase domain 21 pseudogene 1
- Gene Aliases
- ADAM21P
- Chromosome Location
- Chr. 14: 70245753 - 70247801 on Build GRCh38
- UniGene ID
- Hs.712468
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
DQ202398.1 |
- |
1 |
664 |
|
View Full Product Details