Gene Transcripts

Gene Symbol
CFAP61
Entrez Gene ID
26074
Gene Name
cilia and flagella associated protein 61
Gene Aliases
C20orf26, CaM-IP3, dJ1002M8.3, dJ1178H5.4
Chromosome Location
Chr. 20: 20052514 - 20360714 on Build GRCh38
UniGene ID
Hs.729168
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015585.3 NP_056400.3 15 1688
XM_005260688.3 XP_005260745.1 15 1651
XM_005260689.2 XP_005260746.1 15 1651
XM_005260690.3 XP_005260747.1 14 1576
XM_005260693.3 XP_005260750.1 15 1651
XM_005260695.4 XP_005260752.1 10 1962
XM_005260696.4 XP_005260753.1 6 535
XM_005260697.1 XP_005260754.1 15 1651
XM_011529211.1 XP_011527513.1 14 1509
XM_011529212.2 XP_011527514.1 15 1651
XM_017027786.1 XP_016883275.1 15 1591
XM_017027787.1 XP_016883276.1 15 1591
XM_017027789.1 XP_016883278.1 15 1651
XM_017027790.1 XP_016883279.1 15 1651
XM_017027791.1 XP_016883280.1 5 433
XM_017027792.1 XP_016883281.1 6 475
GenBank mRNA AK304174.1 - 15 1591
BC028708.2 - 15 1650 4825941

View Full Product Details