Gene Transcripts
- Gene Symbol
- DNAI1
- Entrez Gene ID
- 27019
- Gene Name
- dynein axonemal intermediate chain 1
- Gene Aliases
- CILD1, DIC1, ICS1, PCD
- Chromosome Location
- Chr. 9: 34458752 - 34520989 on Build GRCh38
- UniGene ID
- Hs.112667
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001281428.1 |
NP_001268357.1
|
18, 19 |
2081 |
|
NM_012144.3 |
NP_036276.1
|
18, 19 |
2069 |
|
XM_006716758.3 |
XP_006716821.1
|
17, 18 |
1712 |
|
XM_011517848.1 |
XP_011516150.1
|
15, 16 |
1823 |
|
XM_017014625.1 |
XP_016870114.1
|
15, 16 |
1811 |
|
GenBank mRNA |
AF091619.1 |
- |
18, 19 |
1986 |
|
AK290696.1 |
- |
18, 19 |
1993 |
|
AK302186.1 |
- |
15, 16 |
1612 |
|
AK314639.1 |
- |
18, 19 |
1981 |
|
BC030583.2 |
- |
18, 19 |
1997 |
4837991 |
View Full Product Details