Gene Transcripts

Gene Symbol
APPL2
Entrez Gene ID
55198
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Gene Aliases
DIP13B
Chromosome Location
Chr. 12: 105173297 - 105236230 on Build GRCh38
UniGene ID
Hs.506603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001251904.1 NP_001238833.1 11 1127
NM_001251905.1 NP_001238834.1 11 1152
NM_018171.3 NP_060641.2 11 1109
XM_006719472.1 XP_006719535.1 11 1046
XM_011538530.2 XP_011536832.1 11 2579
XM_011538531.2 XP_011536833.1 11 2242
XM_011538532.2 XP_011536834.1 11 3568
XM_017019551.1 XP_016875040.1 11 1514
XM_017019552.1 XP_016875041.1 11 2224
XM_017019553.1 XP_016875042.1 12 3822
XM_017019554.1 XP_016875043.1 11 1028
GenBank mRNA AK290562.1 - 11 962
AK293783.1 - 11 901
AK296610.1 - 11 908
AK297100.1 - 11 980
AK310377.1 - 11 897
AY113704.1 - 11 1109
BC033731.1 - 11 1013 5168590

View Full Product Details