Gene Transcripts

Gene Symbol
ACER3
Entrez Gene ID
55331
Gene Name
alkaline ceramidase 3
Gene Aliases
APHC, PHCA
Chromosome Location
Chr. 11: 76860873 - 77026797 on Build GRCh38
UniGene ID
Hs.23862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001300953.1 NP_001287882.1 2, 3 270
NM_001300954.1 NP_001287883.1 3, 4 452
NM_001300955.1 NP_001287884.1 2, 3 328
NM_018367.6 NP_060837.3 3, 4 381
XM_011545151.2 XP_011543453.1 3, 4 347
XM_011545152.2 XP_011543454.1 3, 4 317
XM_011545153.2 XP_011543455.1 3, 4 347
XM_017017987.1 XP_016873476.1 3, 4 366
XM_017017988.1 XP_016873477.1 3, 4 347
GenBank mRNA AF214454.1 - 3, 4 335
AF327353.1 - 3, 4 346
AK002100.1 - 4, 5 441
AK293800.1 - 4, 5 352
AK294978.1 - 2, 3 201
AK295142.1 - 3, 4 369
AK295327.1 - 4, 5 465
AK308856.1 - 4, 5 438
AK315000.1 - 3, 4 347
AK316120.1 - 2, 3 266
BC063034.1 - 4, 5 341
BC073853.1 - 3, 4 298 3867010
BG702017.1 - 3, 4 386
DB461413.1 - 3, 4 372

View Full Product Details