Gene Transcripts

Gene Symbol
ANKH
Entrez Gene ID
56172
Gene Name
ANKH inorganic pyrophosphate transport regulator
Gene Aliases
ANK, CCAL2, CMDJ, CPPDD, HANK, MANK
Chromosome Location
Chr. 5: 14704800 - 14871778 on Build GRCh38
UniGene ID
Hs.156727
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_054027.4 NP_473368.1 6 1032
XM_011514067.1 XP_011512369.1 6 995
XM_017009644.1 XP_016865133.1 6 632
GenBank mRNA AB046801.1 - 6 862
AF274753.1 - 6 769
AK001799.1 - 2 157
AK222631.1 - 730
AK315012.1 - 6 730
AY358503.1 - 6 783
BC009835.2 - 6 957 3927236
BC014526.2 - 6 757 3837372

View Full Product Details