Gene Transcripts

Gene Symbol
CBX8
Entrez Gene ID
57332
Gene Name
chromobox 8
Gene Aliases
PC3, RC1
Chromosome Location
Chr. 17: 79794377 - 79797116 on Build GRCh38
UniGene ID
Hs.387258
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020649.2 NP_065700.1 1, 2 187
GenBank mRNA AF174482.1 - 1, 2 162
AF266479.2 - 1, 2 118
AK074560.1 - 1, 2 187
BC008937.2 - 1, 2 84 2821280
BC009376.2 - 1, 2 140 4121509
BC019289.1 - 1, 2 84 2821280

View Full Product Details