Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163034.1 | NP_001156506.1 | 10 | 2000 | |
NM_020761.2 | NP_065812.1 | 10 | 2000 | ||
GenBank mRNA | AB037724.1 | - | 6 | 682 | |
AB082951.1 | - | 10 | 1195 | ||
AK055912.1 | - | 10 | 1846 | ||
AK295964.1 | - | 9 | 1923 | ||
AK296321.1 | - | 10 | 1995 | ||
AY090663.1 | - | 10 | 1557 | ||
BC136652.1 | - | 10 | 1390 | 9020642 | |
BC136654.1 | - | 10 | 1390 | 9020644 |