Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001035507.2 | NP_001030584.1 | 5 | 817 | |
NM_021831.5 | NP_068603.4 | 5 | 817 | ||
NR_104246.1 | 5 | 817 | |||
XM_005264477.2 | XP_005264534.1 | 5 | 817 | ||
XM_011533011.2 | XP_011531313.1 | 5 | 778 | ||
XM_011533012.2 | XP_011531314.1 | 6 | 983 | ||
XM_011533013.2 | XP_011531315.1 | 6 | 786 | ||
GenBank mRNA | AK023398.1 | - | 5 | 772 | |
AK025492.1 | - | 3 | 339 | ||
AK302091.1 | - | 5 | 748 | ||
AL833844.1 | - | 5 | 759 | ||
BC007415.2 | - | 5 | 759 | 2967512 | |
BC018584.1 | - | 5 | 762 |