Gene Transcripts

Gene Symbol
NOL6
Entrez Gene ID
65083
Gene Name
nucleolar protein 6
Gene Aliases
NRAP, UTP22, bA311H10.1
Chromosome Location
Chr. 9: 33460811 - 33473943 on Build GRCh38
UniGene ID
Hs.493709
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022917.4 NP_075068.2 11 1479
NM_139235.3 NP_631981.2 11 1479
XM_017015044.1 XP_016870533.1 11 1415
GenBank mRNA AF361079.1 - 11 1437
AF361080.1 - 11 1437
AF361081.1 - 11 1437
AK056559.1 - 3 193
AK293969.1 - 10 1282
BC030139.2 - 11 1383 4423152

View Full Product Details