Gene Transcripts

Gene Symbol
BBS10
Entrez Gene ID
79738
Gene Name
Bardet-Biedl syndrome 10
Gene Aliases
C12orf58
Chromosome Location
Chr. 12: 76344486 - 76348442 on Build GRCh38
UniGene ID
Hs.96322
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024685.3 NP_078961.3 2 1122
GenBank mRNA AK304178.1 - 2 1095
BC026355.1 - 2 1107 4817227

View Full Product Details