Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_052941.4 | NP_443173.2 | 10 | 1796 | |
GenBank mRNA | AF288814.1 | - | 10 | 1680 | |
AK131094.1 | - | 10 | 1709 | ||
AK304245.1 | - | 6 | 708 | ||
AK312417.1 | - | 10 | 1788 | ||
AL832576.2 | - | 10 | 1726 | ||
BC070055.1 | - | 10 | 1733 | 30345441 |