Gene Transcripts
- Gene Symbol
- CCDC57
- Entrez Gene ID
- 284001
- Gene Name
- coiled-coil domain containing 57
- Gene Aliases
- Chromosome Location
- Chr. 17: 82101458 - 82212892 on Build GRCh38
- UniGene ID
- Hs.631724
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
XM_011523545.2 |
XP_011521847.2
|
19, 20 |
3163 |
|
XM_011523546.2 |
XP_011521848.1
|
18, 19 |
3495 |
|
XM_011523548.2 |
XP_011521850.1
|
19, 20 |
3191 |
|
XM_011523549.2 |
XP_011521851.1
|
18, 19 |
3005 |
|
XM_011523551.2 |
XP_011521853.1
|
19, 20 |
3204 |
|
XM_011523552.2 |
XP_011521854.1
|
17, 18 |
2563 |
|
XM_011523555.2 |
XP_011521857.1
|
16, 17 |
3563 |
|
XM_011523557.2 |
XP_011521859.1
|
12, 13 |
1927 |
|
XM_017024462.1 |
XP_016879951.1
|
19, 20 |
3129 |
|
XM_017024463.1 |
XP_016879952.1
|
19, 20 |
3154 |
|
XM_017024465.1 |
XP_016879954.1
|
19, 20 |
3155 |
|
XM_017024467.1 |
XP_016879956.1
|
19, 20 |
3320 |
|
XM_017024469.1 |
XP_016879958.1
|
19, 20 |
2983 |
|
XM_017024470.1 |
XP_016879959.1
|
18, 19 |
2784 |
|
XM_017024471.1 |
XP_016879960.1
|
18, 19 |
2960 |
|
XM_017024472.1 |
XP_016879961.1
|
19, 20 |
3181 |
|
XM_017024475.1 |
XP_016879964.1
|
19, 20 |
2890 |
|
XM_017024476.1 |
XP_016879965.1
|
17, 18 |
3066 |
|
XM_017024477.1 |
XP_016879966.1
|
18, 19 |
3234 |
|
XM_017024478.1 |
XP_016879967.1
|
16, 17 |
2697 |
|
GenBank mRNA |
AK074334.1 |
- |
4, 5 |
488 |
|
View Full Product Details