Gene Transcripts

Gene Symbol
WFDC10B
Entrez Gene ID
280664
Gene Name
WAP four-disulfide core domain 10B
Gene Aliases
WAP12
Chromosome Location
Chr. 20: 45684651 - 45708899 on Build GRCh38
UniGene ID
Hs.237392
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172006.2 NP_742003.1 4 372
NM_172131.2 NP_742143.1 3 217
XM_017027824.1 XP_016883313.1 5 444
XM_017027825.1 XP_016883314.1 5 697
GenBank mRNA AF454506.1 - 4 372
AF454507.1 - 3 217
BC121071.1 - 4 370 40119593
BC121072.1 - 4 370 40119597

View Full Product Details