Gene Transcripts

Gene Symbol
LINC00482
Entrez Gene ID
284185
Gene Name
long intergenic non-protein coding RNA 482
Gene Aliases
C17orf55
Chromosome Location
Chr. 17: 81302824 - 81309248 on Build GRCh38
UniGene ID
Hs.631762
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_038080.1 4 1180
GenBank mRNA AK096740.1 - 4 1180
BC108932.1 - 3 1128 40025762
BC108933.1 - 3 1128 40025767

View Full Product Details