Gene Transcripts

Gene Symbol
RILPL1
Entrez Gene ID
353116
Gene Name
Rab interacting lysosomal protein like 1
Gene Aliases
GOSPEL, RLP1
Chromosome Location
Chr. 12: 123471073 - 123533718 on Build GRCh38
UniGene ID
Hs.530315
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319243.1 NP_001306172.1 2 641
NM_001319302.1 NP_001306231.1 2 1027
NM_178314.4 NP_847884.2 2 641
XM_006719372.2 XP_006719435.1 2 635
XM_011538282.2 XP_011536584.1 2 676
GenBank mRNA AK096697.1 - 3 483
BC089444.1 - 2 443 30559974
BX281699.1 - 2 294
BX359997.2 - 2 641

View Full Product Details