Gene Transcripts

Gene Symbol
BCL7B
Entrez Gene ID
9275
Gene Name
BCL tumor suppressor 7B
Gene Aliases
Chromosome Location
Chr. 7: 73536353 - 73557735 on Build GRCh38
UniGene ID
Hs.647051
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197244.1 NP_001184173.1 2 296
NM_001301061.1 NP_001287990.1 3 418
NM_001707.3 NP_001698.2 2 296
NR_036682.1 2 279
GenBank mRNA AJ223979.1 - 2 175
AK290091.1 - 2 204
AL553260.3 - 2 563
BC000956.2 - 2 255 3450946
BC001967.2 - 2 162 3461829
BC009548.1 - 2 206 3897805
BI603803.1 - 2 291
BI837138.1 - 2 267
BQ109412.1 - 2 228
X89985.1 - 2 176

View Full Product Details