Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014507.3 | NP_055322.1 | 3 | 898 | |
NM_173467.4 | NP_775738.3 | 4 | 1116 | ||
NR_046423.1 | 3 | 932 | |||
XM_011530156.2 | XP_011528458.1 | 4 | 730 | ||
GenBank mRNA | AL359401.1 | - | 4 | 1083 | |
AL359403.1 | - | 3 | 865 | ||
BC030985.1 | - | 3 | 850 | ||
BC042195.1 | - | 4 | 1109 | 6052380 | |
CA422238.1 | - | 1 | 282 |