Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242911.1 | NP_001229840.1 | 6 | 1284 | |
NM_001242912.1 | NP_001229841.1 | 7 | 1363 | ||
NM_001242913.1 | NP_001229842.1 | 8 | 1318 | ||
NM_001242914.1 | NP_001229843.1 | 7 | 1293 | ||
NM_001242915.1 | NP_001229844.1 | 6 | 1129 | ||
NM_001242916.1 | NP_001229845.1 | 7 | 1292 | ||
NM_001242917.1 | NP_001229846.1 | 7 | 1292 | ||
NM_001242918.1 | NP_001229847.1 | 5 | 1170 | ||
NM_001242919.1 | NP_001229848.1 | 6 | 1124 | ||
NM_019006.3 | NP_061879.2 | 7 | 1447 | ||
XM_011521684.2 | XP_011519986.1 | 8 | 1330 | ||
GenBank mRNA | AF061739.1 | - | 7 | 1232 | |
AF261138.1 | - | 6 | 1118 | ||
AJ251095.1 | - | 6 | 1080 | ||
AK055881.1 | - | 2 | 2241 | ||
AL136598.1 | - | 6 | 1139 | ||
BC005283.1 | - | 7 | 1336 | 4042180 |