Gene Transcripts
- Gene Symbol
- WDR59
- Entrez Gene ID
- 79726
- Gene Name
- WD repeat domain 59
- Gene Aliases
- FP977
- Chromosome Location
- Chr. 16: 74873568 - 74985146 on Build GRCh38
- UniGene ID
- Hs.280951
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001324171.1 |
NP_001311100.1
|
16, 17 |
1773 |
|
NM_030581.3 |
NP_085058.3
|
16, 17 |
1746 |
|
XM_005256146.3 |
XP_005256203.1
|
16, 17 |
1750 |
|
XM_011523331.2 |
XP_011521633.1
|
14, 15 |
1510 |
|
XM_011523332.2 |
XP_011521634.1
|
10, 11 |
1198 |
|
XM_017023667.1 |
XP_016879156.1
|
15, 16 |
1607 |
|
XM_017023668.1 |
XP_016879157.1
|
14, 15 |
1383 |
|
XM_017023669.1 |
XP_016879158.1
|
14, 15 |
1398 |
|
XM_017023670.1 |
XP_016879159.1
|
13, 14 |
1431 |
|
GenBank mRNA |
AB067510.1 |
- |
11, 12 |
1219 |
|
AF370390.1 |
- |
4, 5 |
678 |
|
AK091316.1 |
- |
4, 5 |
526 |
|
AL832078.1 |
- |
1, 2 |
154 |
|
BC004519.2 |
- |
16, 17 |
1710 |
3938206 |
BC014887.2 |
- |
10, 11 |
1138 |
|
View Full Product Details