Gene Transcripts

Gene Symbol
SCRT2
Entrez Gene ID
85508
Gene Name
scratch family transcriptional repressor 2
Gene Aliases
ZNF898B
Chromosome Location
Chr. 20: 661596 - 676179 on Build GRCh38
UniGene ID
Hs.355284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033129.3 NP_149120.1 2 896
GenBank mRNA AY129025.1 - 2 902

View Full Product Details