Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033549.4 | NP_291027.3 | 1 | 803 | |
NM_201627.2 | NP_963921.1 | 1 | 803 | ||
NR_045218.1 | 1 | 803 | |||
XM_006714929.2 | XP_006714992.1 | 1 | 801 | ||
XM_006714930.2 | XP_006714993.1 | 1 | 801 | ||
XM_006714931.3 | XP_006714994.1 | 1 | 801 | ||
XM_006714932.3 | XP_006714995.1 | 1 | 801 | ||
GenBank mRNA | AB100366.1 | - | 1 | 66 | |
AB100367.1 | - | 1 | 66 | ||
AK027601.1 | - | 1 | 776 | ||
AX775793.1 | - | 1 | 776 | ||
AX775799.1 | - | 1 | 776 | ||
AX775801.1 | - | 1 | 776 | ||
BC090953.1 | - | 1 | 346 | 6017628 | |
BC127195.1 | - | 1 | 590 |