Gene Transcripts

Gene Symbol
TMEM61
Entrez Gene ID
199964
Gene Name
transmembrane protein 61
Gene Aliases
Chromosome Location
Chr. 1: 54980614 - 54992293 on Build GRCh38
UniGene ID
Hs.663950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182532.2 NP_872338.1 2 453
XM_005270586.4 XP_005270643.1 2 91
XM_011540911.2 XP_011539213.1 3 205
XM_011540912.2 XP_011539214.1 2 981
GenBank mRNA BC029775.1 - 2 325
BG430102.1 - 2 453

View Full Product Details