Gene Transcripts
- Gene Symbol
- C6orf1
- Entrez Gene ID
- 221491
- Gene Name
- chromosome 6 open reading frame 1
- Gene Aliases
- LBH
- Chromosome Location
- Chr. 6: 34246380 - 34249108 on Build GRCh38
- UniGene ID
- Hs.381300
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008703.2 |
NP_001008703.1
|
4, 5 |
451 |
|
NM_001008704.2 |
NP_001008704.1
|
4, 5 |
391 |
|
NM_001287396.1 |
NP_001274325.1
|
4, 5 |
643 |
|
NM_001287397.1 |
NP_001274326.1
|
3, 4 |
677 |
|
NM_001287398.1 |
NP_001274327.1
|
3, 4 |
425 |
|
NM_178508.4 |
NP_848603.2
|
4, 5 |
703 |
|
GenBank mRNA |
AA779779.1 |
- |
2, 3 |
417 |
|
AK290164.1 |
- |
4, 5 |
385 |
|
BC023627.2 |
- |
4, 5 |
323 |
4764944 |
BC047919.1 |
- |
4, 5 |
584 |
6190218 |
BM474526.1 |
- |
3, 4 |
342 |
|
BU194178.1 |
- |
4, 5 |
524 |
|
BU956518.1 |
- |
3, 4 |
190 |
|
CK001844.1 |
- |
4, 5 |
454 |
|
View Full Product Details