Gene Transcripts

Gene Symbol
EMC10
Entrez Gene ID
284361
Gene Name
ER membrane protein complex subunit 10
Gene Aliases
C19orf63, HSM1, HSS1
Chromosome Location
Chr. 19: 50476477 - 50505905 on Build GRCh38
UniGene ID
Hs.448941
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175063.5 NP_778233.4 4 435
NM_206538.3 NP_996261.1 4 435
XM_006723163.3 XP_006723226.1 4 429
XM_011526818.2 XP_011525120.1 4 429
GenBank mRNA AY194293.2 - 8 1393
AY358710.1 - 4 395
AY761095.1 - 4 413
AY761097.1 - 4 413
BC032948.2 - 4 376 4794992
BC035001.2 - 4 374 4796476
BC062607.1 - 4 577
BX366174.2 - 4 433
CN423491.1 - 1 60
HY107516.1 - 4 436
HY149334.1 - 1 69

View Full Product Details