Gene Transcripts

Gene Symbol
C7orf71
Entrez Gene ID
285941
Gene Name
chromosome 7 open reading frame 71
Gene Aliases
Chromosome Location
Chr. 7: 26635582 - 26647272 on Build GRCh38
UniGene ID
Hs.413394
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145531.2 NP_001139003.1 3 928
XM_017012010.1 XP_016867499.1 3 3418
XM_017012011.1 XP_016867500.1 4 3304
XM_017012012.1 XP_016867501.1 3 3030
XM_017012013.1 XP_016867502.1 4 2946
GenBank mRNA AK093321.1 - 3 928
BC112220.1 - 2 271
BC112222.1 - 2 272

View Full Product Details