Gene Transcripts

Gene Symbol
C17orf82
Entrez Gene ID
388407
Gene Name
chromosome 17 open reading frame 82
Gene Aliases
Chromosome Location
Chr. 17: 61411751 - 61413280 on Build GRCh38
UniGene ID
Hs.434459
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_203425.2 NP_982249.2 1 599
GenBank mRNA BC046200.1 - 1 599

View Full Product Details