Gene Transcripts

Gene Symbol
C5orf46
Entrez Gene ID
389336
Gene Name
chromosome 5 open reading frame 46
Gene Aliases
SSSP1
Chromosome Location
Chr. 5: 147850463 - 147906594 on Build GRCh38
UniGene ID
Hs.660038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_206966.2 NP_996849.2 1, 2 101
XM_005268446.3 XP_005268503.2 1, 2 148
XM_017009459.1 XP_016864948.1 1, 2 157
GenBank mRNA AK289403.1 - 1, 2 101
AY358656.1 - 1, 2 65
BC021680.1 - 1, 2 105 3931172

View Full Product Details