Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001137601.2 | NP_001131073.1 | 1 | 352 | |
XM_017020911.1 | XP_016876400.1 | 2 | 456 | ||
GenBank mRNA | AX721091.1 | - | 2 | 457 | |
BC157833.1 | - | 1 | 362 | 9057052 | |
BC171822.1 | - | 1 | 362 | 9054476 |