Gene Transcripts

Gene Symbol
KTN1-AS1
Entrez Gene ID
100129075
Gene Name
KTN1 antisense RNA 1
Gene Aliases
C14orf33, MYCLo-3
Chromosome Location
Chr. 14: 55576157 - 55580092 on Build GRCh38
UniGene ID
Hs.633724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027123.1 3 405
GenBank mRNA BX248296.1 - 3 387
JX046911.1 - 3 410
JX628584.1 - 3 410

View Full Product Details