Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001099218.2 | NP_001092688.1 | 3 | 3447 | |
NM_001321233.1 | NP_001308162.1 | 7 | 3805 | ||
XM_011533084.1 | XP_011531386.1 | 2 | 3366 | ||
GenBank mRNA | AK310498.1 | - | 8 | 4052 | |
BG216055.1 | - | 2 | 182 | ||
DB514225.1 | - | 2 | 152 | ||
DQ860102.1 | - | 3 | 3447 |