Gene Transcripts

Gene Symbol
C2orf81
Entrez Gene ID
388963
Gene Name
chromosome 2 open reading frame 81
Gene Aliases
hCG40743
Chromosome Location
Chr. 2: 74414176 - 74421630 on Build GRCh38
UniGene ID
Hs.445377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145054.1 NP_001138526.1 2 535
NM_001316764.1 NP_001303693.1 2 287
NM_001316766.1 NP_001303695.1 2 287
GenBank mRNA BM045416.1 - 2 539
BU184705.1 - 2 248
HY050307.1 - 2 289

View Full Product Details