Gene Transcripts

Gene Symbol
C2orf81
Entrez Gene ID
388963
Gene Name
chromosome 2 open reading frame 81
Gene Aliases
hCG40743
Chromosome Location
Chr. 2: 74414176 - 74421630 on Build GRCh38
UniGene ID
Hs.445377
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001316766.1 NP_001303695.1 2 404
GenBank mRNA BU184705.1 - 2 365

View Full Product Details