Gene Transcripts
- Gene Symbol
- ACTA1
- Entrez Gene ID
- 58
- Gene Name
- actin, alpha 1, skeletal muscle
- Gene Aliases
- ACTA, ASMA, CFTD, CFTD1, CFTDM, MPFD, NEM1, NEM2, NEM3, SHPM
- Chromosome Location
- Chr. 1: 229431245 - 229434096 on Build GRCh38
- UniGene ID
- Hs.1288
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY280960.1 |
- |
2 |
420 |
|
- Gene Symbol
- POTEKP
- Entrez Gene ID
- 440915
- Gene Name
- POTE ankyrin domain family member K, pseudogene
- Gene Aliases
- ACT, ACTBL3, POTE2delta, POTEK
- Chromosome Location
- Chr. 2: 131592093 - 131609401 on Build GRCh38
- UniGene ID
- Hs.631267
- Species
- Homo sapiens
View Full Product Details